5-Chloro-2,4-difluorobenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

PC303353-10G PC303353-1G PC303353-5G
77414-386EA 254.7 USD
77414-386 77414-388 77414-390
5-Chloro-2,4-difluorobenzaldehyde 98%
5-Chloro-2,4-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
MW: 176.548 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782052
CAS Number: 695187-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR