2,5-Difluoro-4-methoxybenzoic acid 97%

Supplier: APOLLO SCIENTIFIC

PC300817-1G PC300817-5G
77414-052EA 87.62 USD
77414-052 77414-054
2,5-Difluoro-4-methoxybenzoic acid 97%
2,5-Difluoro-4-methoxybenzoic acid
Formula: C₈H₆F₂O₃
MW: 188.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847151
CAS Number: 1060739-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR