Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,5-difluorobenzoate

Supplier: APOLLO SCIENTIFIC

PC300790-1G PC300790-250MG PC300790-5G
77414-040EA 308.7 USD
77414-040 77414-042 77414-044
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,5-difluorobenzoate
Methyl 4-(2,6-dimethylmorpholin-4-yl)-2,5-difluorobenzoate
Formula: C₁₄H₁₇F₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD28103351
CAS Number: 1858250-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR