2,2-Difluorosuccinic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α,α-Difluorosuccinic acid, 2,2-Difluoro-1,4-butanedioic acid

PC2890-1G PC2890-5G
77413-966EA 66.22 USD
77413-966 77413-968
2,2-Difluorosuccinic acid 95%
2,2-Difluorosuccinic acid
Formula: C₄H₄F₂O₄
Melting Pt: 144…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00040970
CAS Number: 665-31-6
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR