2-Amino-6-fluorobenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1076C-1G PC1076C-25G PC1076C-5G
77412-968EA 69.28 USD
77412-968 77412-970 77412-972
2-Amino-6-fluorobenzylamine 97%
2-Amino-6-fluorobenzylamine
Formula: C₇H₉FN₂
Boiling Pt: 78…80 °C/1 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042483
CAS Number: 175277-93-7
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR