2,3-Difluoropyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC10418-25G PC10418-5G
77412-888EA 160.98 USD
77412-888 77412-890
2,3-Difluoropyridine 97%
2,3-Difluoropyridine
Formula: C₅H₃F₂N
Storage Temperature: Ambient
MDL Number: MFCD04114143
CAS Number: 1513-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR