α,α,α-Trifluoro-m-toluidine 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride

PC1030-100G PC1030-1KG PC1030-500G
77412-800EA 21.32 USD
77412-800 77412-802 77412-804
α,α,α-Trifluoro-m-toluidine 99%
α,α,α-Trifluoro-m-toluidine

Intermediate for the synthesis of several pharmaceuticals including Bendrofluazide.

Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 187…189 °C
Melting Pt: 44352 °C
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007797
CAS Number: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR