1,1-Difluoroacetone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1013-1G PC1013-25G PC1013-5G
77412-782EA 21.42 USD
77412-782 77412-784 77412-786
1,1-Difluoroacetone 97%
1,1-Difluoroacetone
Formula: C₃H₄F₂O
Boiling Pt: 46.5…46.7 °C/757 mmHg
Density: 1.164 g/cm³ (25 °C)
Storage Temperature: Refrigerator
CAS Number: 431-05-0
UN: 1224
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR