3-Fluoro-4-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Cyano-2-fluorobenzotrifluoride, 3-Fluoro-4-trifluoromethylbenzonitrile

PC0891-100G PC0891-25G PC0891-5G
77412-726EA 172.18 USD
77412-726 77412-728 77412-730
3-Fluoro-4-(trifluoromethyl)benzonitrile 97%
3-Fluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 194 °C
Melting Pt: 45…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00236280
CAS Number: 231953-38-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR