α-Chloro-3,5-difluorotoulene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Chloromethyl)-3,5-difluorobenzene, 3,5-Difluorobenzyl chloride

PC0852-100G PC0852-25G PC0852-5G
77412-702EA 213.74 USD
77412-702 77412-704 77412-706
α-Chloro-3,5-difluorotoulene 98%
α-Chloro-3,5-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 82 °C/22 mmHg
Storage Temperature: Ambient
MDL Number: MFCD01090990
CAS Number: 220141-71-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR