2,2-Difluoroethylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0806-5G PC0806-25G
77412-694EA 22.42 USD
77412-694 77412-692
2,2-Difluoroethylamine 97%
2,2-Difluoroethylamine
Formula: C₂H₅F₂N
Boiling Pt: 68…69 °C
Density: 1.15 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00798137
CAS Number: 430-67-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR