FCCP 95+%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: FCCP, [4-(Trifluoromethoxy)phenyl]carbonohydrazonoyl dicyanide, [4-(Trifluoromethoxy)phenylhydrazono]malononitrile, Carbonyl cyanide 4-(Trifluoromethoxy)phenylhydrazone

PC0578-1G PC0578-250MG
77412-640EA 109.56 USD
77412-640 77412-642 77412-644
FCCP 95+%
FCCP
Formula: C₁₀H₅F₃N₄O
Melting Pt: 174…175 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00009699
CAS Number: 370-86-5
EINECS: 206-730-8
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR