2-Bromo-6-fluoropyridine 97+%

Supplier: APOLLO SCIENTIFIC
Danger

PC3082-100G PC3082-25G PC3082-5G
77414-444EA 144.68 USD
77414-444 77414-446 77414-448
2-Bromo-6-fluoropyridine 97+%
2-Bromo-6-fluoropyridine
Formula: C₅H₃BrFN
Boiling Pt: 162…164 °C
Melting Pt: 30…34 °C
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD03095258
CAS Number: 144100-07-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR