5-Methyl-2-(trifluoromethoxy)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

PC303072-1G PC303072-250MG
77414-354EA 2091.62 USD
77414-354 77414-356
5-Methyl-2-(trifluoromethoxy)benzenesulfonyl chloride 97%
5-Methyl-2-(trifluoromethoxy)benzenesulfonyl chloride
Formula: C₈H₆ClF₃O₃S
Storage Temperature: Ambient
MDL Number: MFCD18399740
CAS Number: 1261683-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR