(2,6-Difluorophenyl)acetonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(2,6-Difluorophenyl)acetonitrile, 2,6-Difluorobenzyl cyanide

PC2874EJ-25G PC2874EJ-5G
77413-946EA 41.78 USD
77413-946 77413-948
(2,6-Difluorophenyl)acetonitrile 98%
(2,6-Difluorophenyl)acetonitrile
Formula: C₈H₅F₂N
MW: 153.13 g/mol
Boiling Pt: 109 °C/27 mmHg
Density: 1.238 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00009973
CAS Number: 654-01-3
EINECS: 211-504-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR