2,6-Difluorophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2870-25G PC2870-5G
77413-936EA 49.92 USD
77413-936 77413-938
2,6-Difluorophenol 98%
2,6-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 59…61 °C/17 mmHg
Melting Pt: 38…41 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002158
CAS Number: 28177-48-2
EINECS: 248-884-9
UN: 2925
ADR: 4.1+8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR