1-Chloro-2,3,3-trifluorocyclobutene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2035-5G PC2035-1G
77413-494EA 269.98 USD
77413-494 77413-492
1-Chloro-2,3,3-trifluorocyclobutene 97%
1-Chloro-2,3,3-trifluorocyclobutene
Formula: C₄H₂ClF₃
MW: 142.51 g/mol
Boiling Pt: 51…52 °C
Density: 1.355 g/cm³ (25 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00041524
CAS Number: 694-62-2
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR