4-Chloro-3-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Chloro-5-hydroxybenzotrifluoride

PC1930L-100G PC1930L-25G PC1930L-5G
77413-414EA 80 USD
77413-414 77413-416 77413-418
4-Chloro-3-(trifluoromethyl)phenol 98%
4-Chloro-3-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
MW: 196.56 g/mol
Boiling Pt: 120…122 °C/27 mmHg
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00042525
CAS Number: 6294-93-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR