(1R)-1-(2-Fluorophenyl)ethylamine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-1-(2-Fluorophenyl)ethanamine

PC0611-100MG PC0611-250MG
77412-660EA 35.4 USD
77412-660 77412-662
(1R)-1-(2-Fluorophenyl)ethylamine
(1R)-1-(2-Fluorophenyl)ethylamine
Formula: C₈H₁₀FN
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03092993
CAS Number: 185545-90-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR