3-Methyl-5-(trifluoromethyl)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC0483-1G PC0483-5G
77412-580EA 329.08 USD
77412-580 77412-582
3-Methyl-5-(trifluoromethyl)benzyl bromide
3-Methyl-5-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃
MW: 253.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631610
CAS Number: 116070-36-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR