4-Fluoro-3-toluoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Fluoro-3-methylbenzoyl chloride, 4-Fluoro-3-methylbenzene-1-carbonyl chloride

PC0350-1G PC0350-25G PC0350-5G
77412-496EA 21.26 USD
77412-496 77412-498 77412-500
4-Fluoro-3-toluoyl chloride 98%
4-Fluoro-3-toluoyl chloride
Formula: C₈H₆ClFO
Boiling Pt: 129…133 °C/17 mmHg
Density: 1.215 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631426
CAS Number: 455-84-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR