1-Chloro-3,3,3-trifluoroacetone

Supplier: APOLLO SCIENTIFIC
Warning

PC0171-25G PC0171-5G
77412-428EA 40.17 USD
77412-428 77412-430 77412-432
1-Chloro-3,3,3-trifluoroacetone
1-Chloro-3,3,3-trifluoroacetone
Formula: C₃H₂ClF₃O
MW: 146.5 g/mol
Boiling Pt: 71…72 °C
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01631508
CAS Number: 431-37-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR