4-Chloro-2-fluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0162-25G PC0162-5G
77412-422EA 38.72 USD
77412-422 77412-424
4-Chloro-2-fluorobenzonitrile 98%
4-Chloro-2-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Boiling Pt: 92 °C/22 mmHg
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00143284
CAS Number: 57381-51-8
EINECS: 260-712-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR