1,3-Difluorobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger

PC2630-100G PC2630-500G
77413-734EA 21.36 USD
77413-734 77413-736
1,3-Difluorobenzene 99%
1,3-Difluorobenzene
Formula: C₆H₄F₂
Boiling Pt: 82 °C
Melting Pt: –59 °C
Density: 1.163 g/cm³ (25 °C)
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00000327
CAS Number: 372-18-9
EINECS: 206-746-5
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR