4-Chloro-3-(trifluoromethyl)benzaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2398-100G PC2398-25G PC2398-5G
77413-616EA 124.14 USD
77413-616 77413-618 77413-620
4-Chloro-3-(trifluoromethyl)benzaldehyde 98%
4-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
Boiling Pt: 54 °C/10 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00040722
CAS Number: 34328-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR