3,3,3-Trifluoro-2-methylpropanoic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC201243-1G PC201243-25G PC201243-5G
77413-468EA 171.16 USD
77413-468 77413-470 77413-472
3,3,3-Trifluoro-2-methylpropanoic acid 97%
3,3,3-Trifluoro-2-methylpropanoic acid
Formula: C₄H₅F₃O₂
MW: 142.077 g/mol
Boiling Pt: 67 °C (5 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD19228247
CAS Number: 381-97-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR