3-Chloro-4-fluorobenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1864F-1G PC1864F-25G PC1864F-5G
77413-382EA 21.26 USD
77413-382 77413-384 77413-386
3-Chloro-4-fluorobenzylamine 97%
3-Chloro-4-fluorobenzylamine
Formula: C₇H₇ClFN
Boiling Pt: 104…106 °C/15 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00042507
CAS Number: 72235-56-4
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR