2,4-Dichloroaniline 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR9687-100G OR9687-250G
77412-274EA 96.78 USD
77412-274 77412-276
2,4-Dichloroaniline 99%
2,4-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 245 °C
Melting Pt: 60…62 °C
Density: 1.567 g/cm³ (25 °C)
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00007661
CAS Number: 554-00-7
EINECS: 209-057-8
UN: 3442
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR