Dibromo(fluoro)acetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,2-Dibromo-2-fluoroethanoic acid

PC2253M-1G PC2253M-250MG PC2253M-5G
77413-576EA 134.48 USD
77413-576 77413-578 77413-580
Dibromo(fluoro)acetic acid 97%
Dibromo(fluoro)acetic acid
Formula: C₂HBr₂FO₂
MW: 235.835 g/mol
Boiling Pt: 130 °C (60 mmHg)
Melting Pt: 40…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00069082
CAS Number: 353-99-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR