4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Bromo-2-(trifluoromethoxy)benzene-1-sulphonyl chloride

PC1593R-100G PC1593R-25G PC1593R-5G
77413-240EA 223.92 USD
77413-240 77413-242 77413-244
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride 97%
4-Bromo-2-(trifluoromethoxy)benzenesulfonylchloride
Formula: C₇H₃BrClF₃O₃S
MW: 339.52 g/mol
Boiling Pt: 255…256 °C
Density: 1.85 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00179336
CAS Number: 175278-14-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR