3-Bromo-4-fluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1420H-100G PC1420H-25G PC1420H-500G
77413-158EA 88.64 USD
77413-158 77413-160 77413-162
3-Bromo-4-fluorobenzonitrile 98%
3-Bromo-4-fluorobenzonitrile
Formula: C₇H₃BrFN
Boiling Pt: 134…136 °C/33 mmHg
Melting Pt: 53…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00055432
CAS Number: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR