4-Amino-3-fluorophenol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-4-hydroxyaniline

PC1076J-100G PC1076J-25G PC1076J-5G
77412-974EA 124.3 USD
77412-974 77412-976 77412-978
4-Amino-3-fluorophenol
4-Amino-3-fluorophenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077452
CAS Number: 399-95-1
EINECS: 402-230-0
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR