2-Bromo-6-fluorobenzonitrile 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC10431-100G
PC10431-25G
PC10431-5G
77412-894EA
140.6
USD
77412-894
77412-896
77412-898
2-Bromo-6-fluorobenzonitrile 98%
2-Bromo-6-fluorobenzonitrile
Formula:
C₇H₃BrFN Melting Pt: 58…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797753 CAS Number: 79544-27-7 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...