Tropylium tetrafluoroborate 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC0915-5G PC0915-25G
77412-734EA 58.08 USD
77412-734 77412-732
Tropylium tetrafluoroborate 95%
Tropylium tetrafluoroborate
Formula: C₇H₇BF₄
Melting Pt: 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00013402
CAS Number: 27081-10-3
EINECS: 248-214-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR