Coumarin-3-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR9923-100G OR9923-25G OR9923-500G
77412-316EA 124.3 USD
77412-316 77412-318 77412-320
Coumarin-3-carboxylic acid 97%
Coumarin-3-carboxylic acid
Formula: C₁₀H₆O₄
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00006852
CAS Number: 531-81-7
EINECS: 208-518-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR