2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC10713-100MG PC10713-1G PC10713-250MG
77412-952EA 47.2 USD
77412-952 77412-954 77412-956
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
Formula: C₇H₅ClF₃NO₂S
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD14583150
CAS Number: 1257535-05-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR