5-Chloro-2-(trifluoromethyl)benzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-5-chloro-2-(trifluoromethyl)benzene, 2-(Bromomethyl)-4-chloro-1-(trifluoromethyl)benzene

PC0195-10G PC0195-1G PC0195-5G
77412-450EA 101.88 USD
77412-450 77412-452 77412-454
5-Chloro-2-(trifluoromethyl)benzyl bromide 98%
5-Chloro-2-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrClF₃
MW: 273.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631356
CAS Number: 261763-24-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR