3-Chloro-2-fluorobenzoyl chloride 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC0044-1G
PC0044-25G
PC0044-5G
77412-354EA
21.26
USD
77412-354
77412-356
77412-358
3-Chloro-2-fluorobenzoyl chloride 97%
3-Chloro-2-fluorobenzoyl chloride
Formula:
C₇H₃Cl₂FO Boiling Pt: 202…203 °C Density: 1.451 g/cm³ (25 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631548 CAS Number: 85345-76-2 UN: 2920 ADR: 8+3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...