6-Bromo-3-pyridazinol 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 6-Bromopyridazin-3(2H)-one

OR943012-10G OR943012-1G OR943012-5G
77412-096EA 323.98 USD
77412-096 77412-098 77412-100
6-Bromo-3-pyridazinol 98%
6-Bromo-3-pyridazinol
Formula: C₄H₃BrN₂O
MW: 174.983 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223475
CAS Number: 51355-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR