5-Bromobenzofuran-2-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR9879-1G OR9879-500MG OR9879-5G
77412-310EA 89.66 USD
77412-310 77412-312 77412-314
5-Bromobenzofuran-2-carboxylic acid
5-Bromobenzofuran-2-carboxylic acid
Formula: C₉H₅BrO₃
MW: 241.0400 g/mol
Melting Pt: 257…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00093973
CAS Number: 10242-11-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR