1-Bromo-3-methoxypropane 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR911631-100G OR911631-25G
77411-616EA 41.78 USD
77411-616 77411-618
1-Bromo-3-methoxypropane 97%
1-Bromo-3-methoxypropane
Formula: C₄H₉BrO
Boiling Pt: 132 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: <60 °C
Storage Temperature: Ambient
MDL Number: MFCD02258473
CAS Number: 36865-41-5
UN: 2344
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR