5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine

Supplier: APOLLO SCIENTIFIC

OR904269-1G OR904269-25G OR904269-5G
77411-546EA 24.46 USD
77411-546 77411-548 77411-550
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
5-(Methoxymethyl)-1,3,4-thiadiazol-2-amine
Formula: C₄H₇N₃OS
MW: 145.1829 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01203002
CAS Number: 15884-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR