cis-1,2-Cyclohexanedicarboxylic acid 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: cis-Cyclohexane-1,2-dicarboxylic acid

OR903673-100G OR903673-500G
77411-536EA 44.82 USD
77411-536 77411-538
cis-1,2-Cyclohexanedicarboxylic acid 98%
cis-1,2-Cyclohexanedicarboxylic acid
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064949
CAS Number: 610-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR