3-(Bromomethyl)phenyl acetate

Supplier: APOLLO SCIENTIFIC
Danger

OR8905-5G OR8905-1G OR8905-25G
77411-474EA 661.22 USD
77411-474 77411-470 77411-472
3-(Bromomethyl)phenyl acetate
3-(Bromomethyl)phenyl acetate
Formula: C₉H₉BrO₂
Storage Temperature: Freezer
MDL Number: MFCD08543447
CAS Number: 49617-80-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR