4-Methylpiperidine

Supplier: APOLLO SCIENTIFIC
Danger

OR8800-100ML OR8800-25ML OR8800-500ML
77411-450EA 34.64 USD
77411-450 77411-452 77411-454
4-Methylpiperidine
4-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 123…124 °C
Melting Pt: 44320 °C
Density: 0.838 g/cm³ (25 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006005
CAS Number: 626-58-4
EINECS: 210-954-1
UN: 2733
ADR: 3+8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR