4-Chloro-2-fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1822-100G PC1822-25G PC1822-500G
77413-326EA 34.64 USD
77413-326 77413-328 77413-330
4-Chloro-2-fluoroaniline 98%
4-Chloro-2-fluoroaniline
Formula: C₆H₅ClFN
Boiling Pt: 92 °C/2 mmHg
Density: 1.311 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00010625
CAS Number: 57946-56-2
EINECS: 261-034-1
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR