4'-Chloro-2'-fluoroacetanilide 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(4-Chloro-2-fluorophenyl)acetamide

PC1815-250G PC1815-50G
77413-316EA 226.64 USD
77413-316 77413-318
4'-Chloro-2'-fluoroacetanilide 98%
4'-Chloro-2'-fluoroacetanilide
Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00042520
CAS Number: 59280-70-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR