3-Methyl-5-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0456-10G PC0456-1G PC0456-5G
77412-568EA 412.62 USD
77412-568 77412-570 77412-572
3-Methyl-5-(trifluoromethyl)benzonitrile 97%
3-Methyl-5-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃N
Storage Temperature: Ambient
MDL Number: MFCD01631595
CAS Number: 261952-04-9
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR