2-Methyl-5-nitrobenzotrifluoride
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
PC0422-100G
PC0422-25G
PC0422-5G
77412-558EA
206.82
USD
77412-558
77412-560
77412-562
2-Methyl-5-nitrobenzotrifluoride
2-Methyl-5-nitrobenzotrifluoride
Formula:
C₈H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631684 CAS Number: 89976-12-5 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...