N-Methyl-N-(trimethylsilyl)trifluoroacetamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: MSTFA, N-Methyl-N-(trimethylsilyl)-2,2,2-trifluoroacetamide, 2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)acetamide

PC0132-250G PC0132-25G PC0132-50G
77412-400EA 166.06 USD
77412-400 77412-402 77412-404
N-Methyl-N-(trimethylsilyl)trifluoroacetamide 98%
N-Methyl-N-(trimethylsilyl)trifluoroacetamide

A powerful silylation reagent with volatile byproducts:J. Chromat., 115, 591 (1975); 134, 387 (1977).

Formula: C₆H₁₂F₃NOSi
MW: 199.25 g/mol
Boiling Pt: 130…132 °C
Density: 1.074 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000411
CAS Number: 24589-78-4
EINECS: 246-331-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR